Search
Search

SCARB1| Gene Symbol: SCARB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001082959.2 | NP_001076428.1 | ||||
| NM_001367981.1 | NP_001354910.1 | |||||
| NM_001367982.1 | NP_001354911.1 | |||||
| NM_001367983.1 | NP_001354912.1 | |||||
| NM_001367984.1 | NP_001354913.1 | |||||
| NM_001367985.1 | NP_001354914.1 | |||||
| NM_001367986.1 | NP_001354915.1 | |||||
| NM_001367987.1 | NP_001354916.1 | |||||
| NM_001367988.1 | NP_001354917.1 | |||||
| NM_001367989.1 | NP_001354918.1 | |||||
| NM_005505.5 | NP_005496.4 | |||||
| NR_160416.1 | ||||||
| NR_160417.1 | ||||||
| NR_160418.1 | ||||||
| NR_160419.1 | ||||||
| NR_160420.1 | ||||||
| NR_160421.1 | ||||||
| NR_160422.1 | ||||||
| NR_160423.1 | ||||||
| NR_160424.1 | ||||||
| GenBank mRNA | AB209436.1 | - | ||||
| AF254409.1 | - | |||||
| AF515445.1 | - | |||||
| AK124112.1 | - | |||||
| AK302779.1 | - | |||||
| AK304730.1 | - | |||||
| BC022087.1 | - | |||||
| BC034322.1 | - | |||||
| BC080647.1 | - | |||||
| BC093732.1 | - | |||||
| BC112037.1 | - | |||||
| BC143319.1 | - | |||||
| BG770204.1 | - | |||||
| CA428516.1 | - | |||||
| KU177964.1 | - | |||||
| KU177965.1 | - | |||||
| Z22555.1 | - | |||||