Search
Search

RIPK1| Gene Symbol: RIPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001317061.1 | NP_001303990.1 | ||||
| NM_003804.4 | NP_003795.2 | |||||
| XM_005249457.3 | XP_005249514.2 | |||||
| XM_006715237.3 | XP_006715300.2 | |||||
| XM_017011403.1 | XP_016866892.1 | |||||
| XM_017011404.1 | XP_016866893.1 | |||||
| XM_017011405.1 | XP_016866894.1 | |||||
| GenBank mRNA | AB208926.1 | - | ||||
| AJ420482.1 | - | |||||
| AK096523.1 | - | |||||
| AK304701.1 | - | |||||
| AK314176.1 | - | |||||
| BC126254.1 | - | 8991970 | ||||
| BC126256.1 | - | 8991972 | ||||
| BX348786.2 | - | |||||
| ENST00000259808.0 | - | |||||
| U25994.1 | - | |||||
| U50062.1 | - | |||||