Search
Search

JUP| Gene Symbol: JUP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001352773.2 | NP_001339702.1 | ||||
| NM_001352774.2 | NP_001339703.1 | |||||
| NM_001352775.2 | NP_001339704.1 | |||||
| NM_001352776.2 | NP_001339705.1 | |||||
| NM_001352777.2 | NP_001339706.1 | |||||
| NM_002230.4 | NP_002221.1 | |||||
| NM_021991.4 | NP_068831.1 | |||||
| XM_006721874.4 | XP_006721937.1 | |||||
| XM_006721875.2 | XP_006721938.1 | |||||
| XM_011524758.2 | XP_011523060.1 | |||||
| XM_017024590.2 | XP_016880079.1 | |||||
| XM_047435934.1 | XP_047291890.1 | |||||
| XM_047435935.1 | XP_047291891.1 | |||||
| XM_047435937.1 | XP_047291893.1 | |||||
| XM_047435938.1 | XP_047291894.1 | |||||
| XM_047435939.1 | XP_047291895.1 | |||||
| XM_047435940.1 | XP_047291896.1 | |||||
| XM_047435941.1 | XP_047291897.1 | |||||
| XM_047435942.1 | XP_047291898.1 | |||||
| GenBank mRNA | AK226051.1 | - | ||||
| AY243535.1 | - | |||||
| BC000441.2 | - | |||||
| BC011865.2 | - | |||||
| KU178155.1 | - | |||||
| KU178156.1 | - | |||||
| M23410.1 | - | |||||
| Z68228.1 | - | |||||