Search
Search

OTUB2| Gene Symbol: OTUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_023112.4 | NP_075601.1 | ||||
| GenBank mRNA | AF318378.1 | - | ||||
| AI018242.1 | - | |||||
| AK025569.1 | - | |||||
| AY177201.1 | - | |||||
| BC000208.1 | - | |||||
| BC009615.1 | - | |||||
| BC068058.1 | - | |||||
| BG423958.1 | - | |||||
| CR457345.1 | - | |||||