Search
Search

CDK11A| Gene Symbol: CDK11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001313896.1 | NP_001300825.1 | ||||
| NM_001313982.1 | NP_001300911.1 | |||||
| NM_024011.3 | NP_076916.2 | |||||
| NM_033529.3 | NP_277071.2 | |||||
| NR_132739.1 | ||||||
| NR_132740.1 | ||||||
| GenBank mRNA | AF067518.1 | - | ||||
| AF067519.1 | - | |||||
| AF067520.1 | - | |||||
| AF067521.1 | - | |||||
| AF067522.1 | - | |||||
| AF067523.1 | - | |||||
| AF067524.1 | - | |||||
| AF067526.1 | - | |||||
| AF067527.1 | - | |||||
| AF067528.1 | - | |||||
| AF067529.1 | - | |||||
| AK301495.1 | - | |||||
| AK303446.1 | - | |||||
| AK303451.1 | - | |||||
| AL043340.1 | - | |||||
| BC014464.1 | - | |||||
| BC028187.1 | - | |||||
| BC073855.1 | - | |||||
| BC094827.1 | - | |||||
| BC110905.1 | - | 6151813 | ||||
| DC388505.1 | - | |||||
| ENST00000407249.0 | - | |||||
| U04819.1 | - | |||||
| U07704.1 | - | |||||
| U07705.1 | - | |||||