Search
Search

Rnf128| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001254761.1 | NP_001241690.1 | ||||
| NM_023270.5 | NP_075759.3 | |||||
| GenBank mRNA | AB041548.1 | - | ||||
| AF426411.1 | - | |||||
| AK004847.1 | - | |||||
| AK008312.1 | - | |||||
| AK018582.1 | - | |||||
| AK146266.1 | - | |||||
| AK167031.1 | - | |||||
| AY112656.1 | - | |||||
| BC010477.1 | - | 3154433 | ||||
| CK380647.1 | - | |||||
| DV064262.1 | - | |||||