Search
Search

| Assay Name: | hsa-miR-146a-5p |
|---|---|
| miRBase Accession Number: | MI0000477 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGAACUGAAUUCCAUGGGUU |
| Species: | Human, Mouse, Rat, Callithrix jacchus, Chicken, Cricetulus griseus, Dog, Horse, Microcebus murinus, Monodelphis domestica, Nomascus leucogenys, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Taeniopygia guttata, Tupaia chinensis |
| Type: | 1 |
| Detection Label: | Alexa Fluor 647 |
| Product Type: | PrimeFlow Probe Set |
Gene Family ID: | MIPF0000103, mir-146 |
Mature miRNA Sequence: | UGAGAACUGAAUUCCAUGGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-146a-5p | MIMAT0000449 |
| Mouse | mmu-miR-146a-5p | MIMAT0000158 |
| Rat | rno-miR-146a-5p | MIMAT0000852 |
| Callithrix jacchus | cja-miR-146a | MIMAT0039478 |
| Chicken | gga-miR-146a-5p | MIMAT0001163 |
| Cricetulus griseus | cgr-miR-146a | MIMAT0023774 |
| View More Rows | ||
| Dog | cfa-miR-146a | MIMAT0006684 |
| Horse | eca-miR-146a | MIMAT0013065 |
| Microcebus murinus | mmr-miR-146a | MIMAT0049129 |
| Monodelphis domestica | mdo-miR-146a-5p | MIMAT0012746 |
| Nomascus leucogenys | nle-miR-146a | MIMAT0049418 |
| Pan troglodytes | ptr-miR-146a | MIMAT0008038 |
| Papio hamadryas | pha-miR-146a | MIMAT0049573 |
| Pig | ssc-miR-146a-5p | MIMAT0022963 |
| Pongo pygmaeus | ppy-miR-146a | MIMAT0015767 |
| Pteropus alecto | pal-miR-146a-5p | MIMAT0040181 |
| Rhesus monkey | mml-miR-146a-5p | MIMAT0006203 |
| Taeniopygia guttata | tgu-miR-146c | MIMAT0014609 |
| Tupaia chinensis | tch-miR-146a-5p | MIMAT0036567 |