Search
Search

| Assay Name: | mmu-miR-3076-5p |
|---|---|
| miRBase Accession Number: | MI0014039 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CACAGGGGAAGCUCAGUGCCAGCC |
| Species: | Mouse |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | CACAGGGGAAGCUCAGUGCCAGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3076-5p | MIMAT0014860 |