Search
Search

| Assay Name: | hsa-miR-6068 |
|---|---|
| miRBase Accession Number: | MI0020345 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUGCGAGUCUCCGGCGGUGG |
| Species: | Human |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | CCUGCGAGUCUCCGGCGGUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-6068 | MIMAT0023693 |