Search
Search

| Assay Name: | hsa-miR-595 |
|---|---|
| miRBase Accession Number: | MI0003607 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAAGUGUGCCGUGGUGUGUCU |
| Species: | Human, Pan troglodytes |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000651, mir-595 |
Mature miRNA Sequence: | GAAGUGUGCCGUGGUGUGUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-595 | MIMAT0003263 |
| Pan troglodytes | ptr-miR-595 | MIMAT0008265 |