Search
Search

| Assay Name: | hsa-miR-199b-5p |
|---|---|
| miRBase Accession Number: | MI0000282 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCCAGUGUUUAGACUAUCUGUUC |
| Species: | Human, Bovine, Dasypus novemcinctus, Goat, Guinea pig, Horse, Monodelphis domestica |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000040, mir-199 |
Mature miRNA Sequence: | CCCAGUGUUUAGACUAUCUGUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-199b-5p | MIMAT0000263 |
| Bovine | bta-miR-199b | MIMAT0003821 |
| Dasypus novemcinctus | dno-miR-199-2-5p | MIMAT0047739 |
| Goat | chi-miR-199b-5p | MIMAT0036038 |
| Goat | chi-miR-199c-5p | MIMAT0036040 |
| Guinea pig | cpo-miR-199-3-5p | MIMAT0047088 |
| View More Rows | ||
| Horse | eca-miR-199b-5p | MIMAT0013780 |
| Monodelphis domestica | mdo-miR-199b-1-5p | MIMAT0004125 |