Search
Search

| Assay Name: | hsa-miR-1272 |
|---|---|
| miRBase Accession Number: | MI0006408 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAUGAUGAUGGCAGCAAAUUCUGAAA |
| Species: | Human, Pan troglodytes |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000645, mir-1272 |
Mature miRNA Sequence: | GAUGAUGAUGGCAGCAAAUUCUGAAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1272 | MIMAT0005925 |
| Pan troglodytes | ptr-miR-1272 | MIMAT0007993 |