Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12499
          Assay Name: hsa-miR-29a-3p
          miRBase Accession Number: MI0000087
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 130876747 - 130876810 [-] on Build GRCh38
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Species: Human, Mouse, Rat, Callithrix jacchus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Papio hamadryas, Pteropus alecto, Rabbit
          Product Type: mirVana® miRNA mimic
          Assay ID MC12499

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000009, mir-29

          Mature miRNA Sequence:

          UAGCACCAUCUGAAAUCGGUUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-29a-3p MIMAT0000086
          Mouse mmu-miR-29a-3p MIMAT0000535
          Rat rno-miR-29a-3p MIMAT0000802
          Callithrix jacchus cja-miR-29a MIMAT0049306
          Dasypus novemcinctus dno-miR-29a-3p MIMAT0047550
          Daubentonia madagascariensis dma-miR-29a MIMAT0049287
          View More Rows
          Dog cfa-miR-29a MIMAT0006626
          Guinea pig cpo-miR-29a-3p MIMAT0046897
          Horse eca-miR-29a MIMAT0012940
          Microcebus murinus mmr-miR-29a MIMAT0049131
          Nomascus leucogenys nle-miR-29a MIMAT0049441
          Otolemur garnettii oga-miR-29a MIMAT0049630
          Papio hamadryas pha-miR-29a MIMAT0049536
          Pteropus alecto pal-miR-29a-3p MIMAT0039802
          Rabbit ocu-miR-29a-3p MIMAT0048144

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-29a
          Stem-loop Accession # MI0000087
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location Chr. 7 - 130876747 - 130876810 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000086
          miRBase ID: hsa-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: Chr. 7 - 130876747 - 130876810 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-29a
          Stem-loop Accession # MI0000576
          Stem-loop Sequence
          ACCCCUUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAGAAUUUUCUAGCACCAUCUGAAAUCGGUUAUAAUGAUUGGGGA
          Chromosome Location Chr. 6 - 31062660 - 31062747 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000535
          miRBase ID: mmu-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: Chr. 6 - 31062660 - 31062747 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-29a
          Stem-loop Accession # MI0000863
          Stem-loop Sequence
          ACCCCUUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAGAAUUUUCUAGCACCAUCUGAAAUCGGUUAUAAUGAUUGGGGA
          Chromosome Location Chr. 4 - 58343931 - 58344018 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000802
          miRBase ID: rno-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: Chr. 4 - 58343931 - 58344018 [-] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-29a
          Stem-loop Accession # MI0040042
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049306
          miRBase ID: cja-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-29a
          Stem-loop Accession # MI0038930
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047550
          miRBase ID: dno-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-29a
          Stem-loop Accession # MI0040018
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049287
          miRBase ID: dma-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-29a
          Stem-loop Accession # MI0008022
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006626
          miRBase ID: cfa-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-29a
          Stem-loop Accession # MI0038586
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046897
          miRBase ID: cpo-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-29a
          Stem-loop Accession # MI0012694
          Stem-loop Sequence
          GAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012940
          miRBase ID: eca-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-29a
          Stem-loop Accession # MI0039843
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049131
          miRBase ID: mmr-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-29a
          Stem-loop Accession # MI0040188
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049441
          miRBase ID: nle-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-29a
          Stem-loop Accession # MI0040412
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049630
          miRBase ID: oga-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-29a
          Stem-loop Accession # MI0040303
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049536
          miRBase ID: pha-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-29a
          Stem-loop Accession # MI0032336
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUACAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039802
          miRBase ID: pal-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-29a
          Stem-loop Accession # MI0039243
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAGAAUUUUCUAGCACCAUCUGAAAUCGGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048144
          miRBase ID: ocu-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464529_pri
          TaqMan™ Pri-miRNA Assay : Mm03306859_pri
          TaqMan™ Pri-miRNA Assay : Hs03302672_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12499
          Ambion® Pre-miR™ miRNA Precursor : PM12499
          TaqMan™ MicroRNA Assay : 002112
          mirVana® miRNA inhibitor : MH12499
          TaqMan™ Advanced miRNA Assay : 478587_mir


          miRBase Alias:

          Human: hsa-miR-29a (v17)
          Mouse: mmu-miR-29a (v17)
          Rat: rno-miR-29a (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline