Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12962
          Assay Name: hsa-miR-129-1-3p
          miRBase Accession Number: MI0000252
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 128207872 - 128207943 [+] on Build GRCh38
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Species: Human, Mouse, Alligator mississippiensis, Dasypus novemcinctus, Guinea pig, Horse, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Salmo salar, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC12962

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000073, mir-129

          Mature miRNA Sequence:

          AAGCCCUUACCCCAAAAAGUAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-129-1-3p MIMAT0004548
          Mouse mmu-miR-129-1-3p MIMAT0016994
          Alligator mississippiensis ami-miR-129a-3p MIMAT0038137
          Dasypus novemcinctus dno-miR-129-3p MIMAT0047625
          Guinea pig cpo-miR-129-3p MIMAT0046978
          Horse eca-miR-129a-3p MIMAT0012933
          View More Rows
          Pongo pygmaeus ppy-miR-129-1-3p MIMAT0015752
          Pteropus alecto pal-miR-129b-3p MIMAT0039873
          Rabbit ocu-miR-129-3p MIMAT0048215
          Rhesus monkey mml-miR-129-1-3p MIMAT0006186
          Salmo salar ssa-miR-129-3-3p MIMAT0032320
          Zebrafish dre-miR-129-3-3p MIMAT0048649

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-129-1
          Stem-loop Accession # MI0000252
          Stem-loop Sequence
          GGAUCUUUUUGCGGUCUGGGCUUGCUGUUCCUCUCAACAGUAGUCAGGAAGCCCUUACCCCAAAAAGUAUCU
          Chromosome Location Chr. 7 - 128207872 - 128207943 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004548
          miRBase ID: hsa-miR-129-1-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: Chr. 7 - 128207872 - 128207943 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-129-1
          Stem-loop Accession # MI0000222
          Stem-loop Sequence
          UGGAUCUUUUUGCGGUCUGGGCUUGCUGUUCUCUCGACAGUAGUCAGGAAGCCCUUACCCCAAAAAGUAUCUA
          Chromosome Location Chr. 6 - 29022619 - 29022691 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0016994
          miRBase ID: mmu-miR-129-1-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: Chr. 6 - 29022619 - 29022691 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-129a
          Stem-loop Accession # MI0029727
          Stem-loop Sequence
          GUCCCUCUCAGAUCUUUUUGCGGUCUGGGCUUGCUGUUCCUCCAACCCACACUCAGGAAGCCCUUACCCCAAAAAGUAUCCGCGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038137
          miRBase ID: ami-miR-129a-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-129-1
          Stem-loop Accession # MI0038972
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUUCUCUCAACAGUAGUCAGGAAGCCCUUACCCCAAAAAGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047625
          miRBase ID: dno-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-129-1
          Stem-loop Accession # MI0038631
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUUUCUUGCAACGGUAGUCAGGAAGCCCUUACCCCAAAAAGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046978
          miRBase ID: cpo-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-129a
          Stem-loop Accession # MI0012687
          Stem-loop Sequence
          UGGAUCUUUUUGCGGUCUGGGCUUGCUGUUCCUCUCAACAGUAGUCAGGAAGCCCUUACCCCAAAAAGUAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012933
          miRBase ID: eca-miR-129a-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-129-1
          Stem-loop Accession # MI0014820
          Stem-loop Sequence
          GGAUCUUUUUGCGGUCUGGGCUUGCUGUUCCUCUCAACAGUAGUCAGGAAGCCCUUACCCCAAAAAGUAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015752
          miRBase ID: ppy-miR-129-1-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-129b
          Stem-loop Accession # MI0032374
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUUCCUCUCAACGAUAGUCAGGAAGCCCUUACCCCAAAAAGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039873
          miRBase ID: pal-miR-129b-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-129-1
          Stem-loop Accession # MI0039283
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUUCCUCUCAGCAGUAGUCAGGAAGCCCUUACCCCAAAAAGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048215
          miRBase ID: ocu-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-129-1
          Stem-loop Accession # MI0007618
          Stem-loop Sequence
          GGAUCUUUUUGCGGUCUGGGCUUGCUGUUCCUCUCAACAGUAGUCAGGAAGCCCUUACCCCAAAAAGUAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006186
          miRBase ID: mml-miR-129-1-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-129-3
          Stem-loop Accession # MI0026467
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUUCAUAAGGCAGUAGCCAGGAAGCCCUUACCCCAAAAAGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032320
          miRBase ID: ssa-miR-129-3-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-129-3
          Stem-loop Accession # MI0039507
          Stem-loop Sequence
          GUCCUUCACGGCUCUUUUUGCGGUCUGGGCUUGCUGUUCCUUAAAUCACCAGUCAGGAAGCCCUUACCCCAAAAAGUAUCUGUGUGGGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048649
          miRBase ID: dre-miR-129-3-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGUAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302824_pri
          TaqMan™ Pri-miRNA Assay : Mm03306410_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12962
          Ambion® Pre-miR™ miRNA Precursor : PM12962
          TaqMan™ MicroRNA Assay : 002298
          mirVana® miRNA inhibitor : MH12962
          TaqMan™ Advanced miRNA Assay : mmu480873_mir
          TaqMan™ Advanced miRNA Assay : 480873_mir


          miRBase Alias:

          Human: hsa-miR-129* (v17)
          Macaca mulatta: mml-miR-129-3p (v19)
          Pongo pygmaeus: ppy-miR-129-1* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline