Search
Search

| Assay Name: | hsa-miR-129-1-3p |
|---|---|
| miRBase Accession Number: | MI0000252 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGCCCUUACCCCAAAAAGUAU |
| Species: | Human, Mouse, Alligator mississippiensis, Dasypus novemcinctus, Guinea pig, Horse, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Salmo salar, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000073, mir-129 |
Mature miRNA Sequence: | AAGCCCUUACCCCAAAAAGUAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-129-1-3p | MIMAT0004548 |
| Mouse | mmu-miR-129-1-3p | MIMAT0016994 |
| Alligator mississippiensis | ami-miR-129a-3p | MIMAT0038137 |
| Dasypus novemcinctus | dno-miR-129-3p | MIMAT0047625 |
| Guinea pig | cpo-miR-129-3p | MIMAT0046978 |
| Horse | eca-miR-129a-3p | MIMAT0012933 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-129-1-3p | MIMAT0015752 |
| Pteropus alecto | pal-miR-129b-3p | MIMAT0039873 |
| Rabbit | ocu-miR-129-3p | MIMAT0048215 |
| Rhesus monkey | mml-miR-129-1-3p | MIMAT0006186 |
| Salmo salar | ssa-miR-129-3-3p | MIMAT0032320 |
| Zebrafish | dre-miR-129-3-3p | MIMAT0048649 |