Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH13108
          Assay Name: hsa-miR-450b-5p
          miRBase Accession Number: MI0005531
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134540185 - 134540262 [-] on Build GRCh38
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Species: Human, Bovine, Gorilla gorilla, Horse, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH13108

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000128, mir-450

          Mature miRNA Sequence:

          UUUUGCAAUAUGUUCCUGAAUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-450b-5p MIMAT0004909
          Bovine bta-miR-450b MIMAT0024576
          Gorilla gorilla ggo-miR-450b MIMAT0024296
          Horse eca-miR-450b-5p MIMAT0013220
          Pig ssc-miR-450b-5p MIMAT0013927
          Pongo pygmaeus ppy-miR-450b-5p MIMAT0015894
          View More Rows
          Rabbit ocu-miR-450b-5p MIMAT0048460
          Rhesus monkey mml-miR-450b-5p MIMAT0006333

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-450b
          Stem-loop Accession # MI0005531
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location Chr. X - 134540185 - 134540262 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004909
          miRBase ID: hsa-miR-450b-5p
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Chromosome Location: Chr. X - 134540185 - 134540262 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-450b
          Stem-loop Accession # MI0021121
          Stem-loop Sequence
          AAAAUUAUUUUUGCAAUAUGUUCCUGAAUAUUUAAUAUAAGUAUAUUGGGAACAUUUUGCAUCCAUAGUUUUGUAUCAGUAUUUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024576
          miRBase ID: bta-miR-450b
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-450b
          Stem-loop Accession # MI0020844
          Stem-loop Sequence
          AAUGUAAGAUGCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAUCAACAUAUGGAGAAAAGUCUAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024296
          miRBase ID: ggo-miR-450b
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-450b
          Stem-loop Accession # MI0012966
          Stem-loop Sequence
          UACAAAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAACAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013220
          miRBase ID: eca-miR-450b-5p
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-450b
          Stem-loop Accession # MI0013140
          Stem-loop Sequence
          AAAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUAUAUUGGGAACAUUUUGCAUCCAUAGUUUUGUAUCGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013927
          miRBase ID: ssc-miR-450b-5p
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-450b
          Stem-loop Accession # MI0014951
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015894
          miRBase ID: ppy-miR-450b-5p
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-450b
          Stem-loop Accession # MI0039412
          Stem-loop Sequence
          UUUUGCAAUAUGUUCCUGAAUAUGUAAUCUAAGUGUAUUGGGGACAUUUUGCAUCCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048460
          miRBase ID: ocu-miR-450b-5p
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-450b
          Stem-loop Accession # MI0007751
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAGUAUAAGCGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006333
          miRBase ID: mml-miR-450b-5p
          Mature miRNA Sequence: UUUUGCAAUAUGUUCCUGAAUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303731_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13108
          Ambion® Pre-miR™ miRNA Precursor : PM13108
          TaqMan™ MicroRNA Assay : 002207
          mirVana® miRNA mimic : MC13108
          TaqMan™ Advanced miRNA Assay : 478914_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline