Search
Search

| Assay Name: | hsa-miR-450b-5p |
|---|---|
| Stem-loop Accession Number: | MI0005531 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUUGCAAUAUGUUCCUGAAUA |
| Species: | Human, Bovine, Gorilla gorilla, Horse, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000128, mir-450 |
Mature miRNA Sequence: | UUUUGCAAUAUGUUCCUGAAUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-450b-5p | MIMAT0004909 |
| Bovine | bta-miR-450b | MIMAT0024576 |
| Gorilla gorilla | ggo-miR-450b | MIMAT0024296 |
| Horse | eca-miR-450b-5p | MIMAT0013220 |
| Pig | ssc-miR-450b-5p | MIMAT0013927 |
| Pongo pygmaeus | ppy-miR-450b-5p | MIMAT0015894 |
| View More Rows | ||
| Rabbit | ocu-miR-450b-5p | MIMAT0048460 |
| Rhesus monkey | mml-miR-450b-5p | MIMAT0006333 |