Search
Search

| Assay Name: | hsa-miR-550a-5p |
|---|---|
| miRBase Accession Number: | MI0003600 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGUGCCUGAGGGAGUAAGAGCCC |
| Species: | Human, Pan troglodytes |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000334, mir-550 |
Mature miRNA Sequence: | AGUGCCUGAGGGAGUAAGAGCCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-550a-5p | MIMAT0004800 |
| Pan troglodytes | ptr-miR-550 | MIMAT0008230 |