Search
Search

Coro2a| Gene Symbol: Coro2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164804.1 | NP_001158276.1 | 2 | 188 | ||
| NM_178893.4 | NP_849224.2 | 3 | 413 | |||
| GenBank mRNA | AK030097.1 | - | 3 | 416 | ||
| AK030971.1 | - | 2 | 144 | |||
| AK044060.1 | - | 2 | 150 | |||
| AK054394.1 | - | 2 | 189 | |||
| AK152490.1 | - | 2 | 177 | |||
| AK153134.1 | - | 2 | 177 | |||
| AK154247.1 | - | 2 | 145 | |||
| AK170184.1 | - | 2 | 146 | |||
| BC138731.1 | - | 2 | 213 | 8861753 | ||
| BC138732.1 | - | 2 | 213 | 8861754 | ||