Search
Search

CPA3| Gene Symbol: CPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001870.3 | NP_001861.2 | 8 | 761 | ||
| GenBank mRNA | AK313628.1 | - | 8 | 721 | ||
| BC012613.1 | - | 8 | 724 | 4104766 | ||
| M27717.1 | - | 8 | 709 | |||
| S40234.1 | - | 5 | 382 | |||