Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NLRP3 SIRNA Assays ›
          Gene Symbol:
          NLRP3
          Gene Name:
          NLR family pyrin domain containing 3
          Gene Aliases:
          AGTAVPRL, AII, AVP, C1orf7, CIAS1, CLR1.1, FCAS, FCAS1, FCU, MWS, NALP3, PYPAF1
          Chromosome Location:
          Chr.1: 247416156 - 247449108 on Build GRCh38
          UniGene ID:
          Hs.159483
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107575

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NLRP3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079821.2 NP_001073289.1 9, 10 2978
          NM_001127461.2 NP_001120933.1 7, 8 3449
          NM_001127462.2 NP_001120934.1 6, 7 3415
          NM_001243133.1 NP_001230062.1 7, 8 3586
          NM_004895.4 NP_004886.3 7, 8 3586
          NM_183395.2 NP_899632.1 5, 6 3244
          XM_011544048.2 XP_011542350.1 8, 9 3725
          XM_011544053.2 XP_011542355.1 6, 7 3271
          XM_011544055.2 XP_011542357.1 5, 6 3100
          XM_017000181.1 XP_016855670.1 9, 10 3202
          XM_017000182.1 XP_016855671.1 9, 10 3147
          XM_017000183.1 XP_016855672.1 7, 8 3442
          XM_017000184.1 XP_016855673.1 6, 7 3271
          GenBank mRNA AB208891.1 - 6, 7 3424
          AF054176.1 - 3, 4 1320
          AF410477.1 - 7, 8 3586
          AF420469.1 - 8, 9 2972
          AF427617.1 - 5, 6 3244
          AF468522.1 - 7, 8 3002
          AK314998.1 - 9, 10 2978
          AY092033.1 - 5, 6 2660
          AY422168.1 - 6, 7 2663
          BC117211.1 - 6, 7 2734 40125762
          BC143359.1 - 8, 9 2845 9051864
          BC143362.1 - 7, 8 2905 9051867
          BC143363.1 - 6, 7 2734 9051868

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of acute inflammatory response
          positive regulation of type 2 immune response
          transcription, DNA-templated
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          defense response
          inflammatory response
          signal transduction
          detection of biotic stimulus
          negative regulation of NF-kappaB transcription factor activity
          interleukin-1 beta production
          interleukin-18 production
          positive regulation of interleukin-13 production
          positive regulation of interleukin-4 production
          positive regulation of interleukin-5 production
          negative regulation of NF-kappaB import into nucleus
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          NLRP3 inflammasome complex assembly
          innate immune response
          positive regulation of T-helper 2 cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          interleukin-1 secretion
          negative regulation of interleukin-1 beta secretion
          positive regulation of interleukin-1 beta secretion
          negative regulation of inflammatory response
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          defense response to virus
          cellular response to lipopolysaccharide
          positive regulation of T-helper 17 cell differentiation
          positive regulation of T-helper 2 cell cytokine production
          protein binding
          ATP binding
          transcription factor binding
          peptidoglycan binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-phcsm:80/100.66.77.16:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0