Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CADM1 SIRNA Assays ›
          Gene Symbol:
          CADM1
          Gene Name:
          cell adhesion molecule 1
          Gene Aliases:
          BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
          Chromosome Location:
          Chr.11: 115173625 - 115504523 on Build GRCh38
          UniGene ID:
          Hs.370510
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 108519

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CADM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098517.1 NP_001091987.1 2, 3 397
          NM_001301043.1 NP_001287972.1 2, 3 397
          NM_001301044.1 NP_001287973.1 2, 3 397
          NM_001301045.1 NP_001287974.1 2, 3 397
          NM_014333.3 NP_055148.3 2, 3 397
          XM_005271494.2 XP_005271551.1 2, 3 290
          XM_017017457.1 XP_016872946.1 2, 3 356
          XM_017017458.1 XP_016872947.1 2, 3 163
          XM_017017459.1 XP_016872948.1 3, 4 2350
          XM_017017460.1 XP_016872949.1 3, 4 2350
          XM_017017461.1 XP_016872950.1 3, 4 364
          GenBank mRNA AB094146.1 - 2, 3 268
          AF132811.1 - 2, 3 271
          AK075502.1 - 2, 3 287
          AK315245.1 - 2, 3 330
          AY358334.1 - 2, 3 283
          BC035930.1 - 2, 3 289
          BC047021.1 - 2, 3 397 5248100
          BC125102.1 - 2, 3 272
          BC125103.1 - 2, 3 271 40118330
          BI915307.1 - 2, 3 397
          CR457157.1 - 2, 3 268
          HE586496.1 - 2, 3 271
          HE586497.1 - 2, 3 271
          HE586498.1 - 2, 3 271
          HE586499.1 - 2, 3 271
          HE586500.1 - 3, 4 357
          HE586503.1 - 3, 4 299
          KJ534791.1 - 2, 3 268
          KJ534792.1 - 2, 3 268
          KJ534793.1 - 2, 3 268
          KJ534794.1 - 2, 3 268
          KJ534795.1 - 2, 3 268
          KJ534796.1 - 2, 3 268
          KJ534797.1 - 2, 3 268

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001012201, XM_006242946, XM_006242944, XM_006242947, XM_006242948, and others.
          This siRNA also targets the following transcripts in Mouse: NM_207675, NM_018770, NM_001025600, NM_207676, AB183401, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell mediated cytotoxicity
          apoptotic process
          homophilic cell adhesion via plasma membrane adhesion molecules
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          multicellular organism development
          spermatogenesis
          cell recognition
          cell differentiation
          adherens junction organization
          susceptibility to natural killer cell mediated cytotoxicity
          positive regulation of natural killer cell mediated cytotoxicity
          positive regulation of cytokine secretion
          activated T cell proliferation
          detection of stimulus
          receptor activity
          receptor binding
          protein binding
          PDZ domain binding
          protein homodimerization activity
          cell adhesion molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0