siRNA Details
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000194.2 | NP_000185.1 | 9 | 1090 | ||
GenBank mRNA | BC000578.2 | - | 9 | 962 | 3163726 | |
M31642.1 | - | 9 | 1008 |

Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000194.2 | NP_000185.1 | 9 | 1090 | ||
GenBank mRNA | BC000578.2 | - | 9 | 962 | 3163726 | |
M31642.1 | - | 9 | 1008 |