Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRMT2 SIRNA Assays ›
          Gene Symbol:
          PRMT2
          Gene Name:
          protein arginine methyltransferase 2
          Gene Aliases:
          HRMT1L1
          Chromosome Location:
          Chr.21: 46635153 - 46665685 on Build GRCh38
          UniGene ID:
          Hs.154163
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 110965

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRMT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242864.2 NP_001229793.1 4, 5 547
          NM_001242865.2 NP_001229794.1 4, 5 547
          NM_001242866.2 NP_001229795.1 4, 5 547
          NM_001286676.1 NP_001273605.1 4, 5 547
          NM_001286677.1 NP_001273606.1 4, 5 547
          NM_001286678.1 NP_001273607.1 4, 5 547
          NM_001535.4 NP_001526.2 4, 5 547
          NM_206962.3 NP_996845.1 5, 6 656
          XM_005261111.3 XP_005261168.1 5, 6 1093
          XM_006723998.3 XP_006724061.1 5, 6 656
          XM_006723999.2 XP_006724062.1 5, 6 656
          XM_006724000.2 XP_006724063.1 5, 6 656
          XM_017028339.1 XP_016883828.1 5, 6 656
          XM_017028340.1 XP_016883829.1 4, 5 989
          GenBank mRNA AF059319.1 - 2, 3 284
          AK123352.1 - 4, 5 1276
          AK123650.1 - 5, 6 1084
          AK223157.1 - 4, 5 468
          AK312573.1 - 4, 5 484
          AY786414.2 - 4, 5 492
          BC000727.2 - 5, 6 576 3506339
          BC038765.1 - 2, 3 535
          BC100026.1 - 4, 5 461 30559810
          BF683143.1 - 4, 5 528
          BQ421001.1 - 5, 6 561
          CR541804.1 - 3, 4 322
          FJ436410.1 - 4, 5 498
          FJ436411.1 - 4, 5 498
          FJ436412.1 - 4, 5 498
          KU178127.1 - 3, 4 322
          KU178128.1 - 3, 4 322
          U79286.1 - 4, 5 489
          U80213.1 - 4, 5 487
          X99209.1 - 4, 5 498

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: AK077456, BC122563, AK144412, AF169620, AK159186, and others.

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein methylation
          signal transduction
          histone methylation
          peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
          negative regulation of NF-kappaB transcription factor activity
          histone arginine methylation
          positive regulation of apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          developmental cell growth
          regulation of androgen receptor signaling pathway
          negative regulation of G1/S transition of mitotic cell cycle
          transcription coactivator activity
          signal transducer activity
          protein binding
          histone-arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          estrogen receptor binding
          progesterone receptor binding
          protein-arginine omega-N asymmetric methyltransferase activity
          histone methyltransferase activity
          protein homodimerization activity
          retinoic acid receptor binding
          peroxisome proliferator activated receptor binding
          thyroid hormone receptor binding
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0