Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ID2 SIRNA Assays ›
Gene Symbol:
ID2
Gene Name:
inhibitor of DNA binding 2, HLH protein
Gene Aliases:
GIG8, ID2A, ID2H, bHLHb26
Chromosome Location:
Chr.2: 8681983 - 8684453 on Build GRCh38
UniGene ID:
Hs.180919
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
11259
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ID2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002166.4
NP_002157.2
3
702
GenBank mRNA
BC030639.2
-
3
629
4820416
D13891.1
-
3
615
M97796.1
-
3
627
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
metanephros development
natural killer cell differentiation
thigmotaxis
membranous septum morphogenesis
bundle of His development
transcription, DNA-templated
multicellular organism development
adult locomotory behavior
positive regulation of gene expression
negative regulation of gene expression
oligodendrocyte development
regulation of lipid metabolic process
olfactory bulb development
circadian regulation of gene expression
mammary gland epithelial cell proliferation
regulation of circadian rhythm
entrainment of circadian clock by photoperiod
enucleate erythrocyte differentiation
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
locomotor rhythm
negative regulation of B cell differentiation
positive regulation of fat cell differentiation
positive regulation of erythrocyte differentiation
positive regulation of macrophage differentiation
negative regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of blood pressure
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
cell maturation
Peyer's patch development
embryonic digestive tract morphogenesis
positive regulation of smooth muscle cell proliferation
neuron fate commitment
cell morphogenesis involved in neuron differentiation
positive regulation of astrocyte differentiation
negative regulation of oligodendrocyte differentiation
adipose tissue development
mammary gland alveolus development
epithelial cell differentiation involved in mammary gland alveolus development
endodermal digestive tract morphogenesis
positive regulation of cell cycle arrest
cellular response to lithium ion
positive regulation of transcription involved in G1/S transition of mitotic cell cycle
cellular senescence
regulation of G1/S transition of mitotic cell cycle
negative regulation of neural precursor cell proliferation
protein binding
ion channel binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0