Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK2 SIRNA Assays ›
Gene Symbol:
ANK2
Gene Name:
ankyrin 2, neuronal
Gene Aliases:
ANK-2, LQT4, brank-2
Chromosome Location:
Chr.4: 112706083 - 113383740 on Build GRCh38
UniGene ID:
Hs.620557
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
112747
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127493.1
NP_001120965.1
33
3968
NM_001148.4
NP_001139.3
31
3874
NM_020977.3
NP_066187.2
31
3874
XM_005262945.2
XP_005263002.1
30
3873
XM_017008067.1
XP_016863556.1
33
4831
XM_017008068.1
XP_016863557.1
34
5072
XM_017008069.1
XP_016863558.1
32
4806
XM_017008070.1
XP_016863559.1
33
5017
XM_017008071.1
XP_016863560.1
34
4000
XM_017008072.1
XP_016863561.1
33
3976
XM_017008073.1
XP_016863562.1
31
3972
XM_017008074.1
XP_016863563.1
33
3946
XM_017008075.1
XP_016863564.1
32
3910
XM_017008076.1
XP_016863565.1
31
3972
XM_017008077.1
XP_016863566.1
30
3787
XM_017008078.1
XP_016863567.1
31
3972
XM_017008079.1
XP_016863568.1
29
3702
XM_017008080.1
XP_016863569.1
31
3823
XM_017008081.1
XP_016863570.1
30
3873
XM_017008082.1
XP_016863571.1
32
4050
XM_017008083.1
XP_016863572.1
32
4113
XM_017008084.1
XP_016863573.1
32
4008
XM_017008085.1
XP_016863574.1
32
3969
XM_017008086.1
XP_016863575.1
31
3870
XM_017008087.1
XP_016863576.1
33
4149
XM_017008088.1
XP_016863577.1
34
3943
XM_017008089.1
XP_016863578.1
31
4116
XM_017008090.1
XP_016863579.1
32
4113
XM_017008091.1
XP_016863580.1
30
3810
XM_017008092.1
XP_016863581.1
32
3910
XM_017008093.1
XP_016863582.1
33
3907
XM_017008094.1
XP_016863583.1
32
3969
XM_017008095.1
XP_016863584.1
30
3787
XM_017008096.1
XP_016863585.1
34
3943
XM_017008097.1
XP_016863586.1
31
3748
XM_017008098.1
XP_016863587.1
32
3910
XM_017008099.1
XP_016863588.1
33
3907
XM_017008100.1
XP_016863589.1
30
3712
XM_017008101.1
XP_016863590.1
30
3873
XM_017008102.1
XP_016863591.1
29
3688
XM_017008103.1
XP_016863592.1
32
3969
XM_017008104.1
XP_016863593.1
31
3811
XM_017008105.1
XP_016863594.1
30
3787
XM_017008106.1
XP_016863595.1
29
3774
XM_017008107.1
XP_016863596.1
31
3748
XM_017008108.1
XP_016863597.1
29
3672
XM_017008109.1
XP_016863598.1
30
3772
XM_017008110.1
XP_016863599.1
31
3811
XM_017008111.1
XP_016863600.1
30
3787
XM_017008112.1
XP_016863601.1
28
3589
GenBank mRNA
AK294720.1
-
10
1619
AK299767.1
-
32
3898
AK299893.1
-
4
716
AK307601.1
-
31
3825
BX537758.1
-
33
3942
BX538132.1
-
5
811
X56958.2
-
31
3789
Z26634.2
-
31
3774
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
atrial septum development
cellular calcium ion homeostasis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
positive regulation of gene expression
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of cardiac muscle contraction by calcium ion signaling
paranodal junction assembly
regulation of protein stability
T-tubule organization
protein localization to organelle
protein localization to cell surface
cellular protein localization
protein localization to M-band
protein localization to T-tubule
positive regulation of potassium ion transport
protein stabilization
regulation of release of sequestered calcium ion into cytosol
response to methylmercury
regulation of calcium ion transport
positive regulation of calcium ion transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
sarcoplasmic reticulum calcium ion transport
protein localization to endoplasmic reticulum
protein localization to plasma membrane
protein targeting to plasma membrane
regulation of cardiac muscle cell contraction
ventricular cardiac muscle cell action potential
atrial cardiac muscle cell action potential
SA node cell action potential
membrane depolarization during SA node cell action potential
atrial cardiac muscle cell to AV node cell communication
SA node cell to atrial cardiac muscle cell communication
regulation of heart rate by cardiac conduction
regulation of SA node cell action potential
regulation of atrial cardiac muscle cell action potential
positive regulation of potassium ion transmembrane transporter activity
regulation of calcium ion transmembrane transporter activity
positive regulation of calcium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
protein kinase binding
spectrin binding
protein binding, bridging
ion channel binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0