Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCF12 SIRNA Assays ›
          Gene Symbol:
          TCF12
          Gene Name:
          transcription factor 12
          Gene Aliases:
          CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
          Chromosome Location:
          Chr.15: 56918090 - 57291261 on Build GRCh38
          UniGene ID:
          Hs.511504
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 115166

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCF12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306219.2 NP_001293148.1 2 239
          NM_001306220.2 NP_001293149.1 2 239
          NM_001322151.1 NP_001309080.1 9 946
          NM_001322152.1 NP_001309081.1 9 951
          NM_001322154.1 NP_001309083.1 9 946
          NM_001322156.1 NP_001309085.1 10 1013
          NM_001322157.1 NP_001309086.1 9 951
          NM_001322158.1 NP_001309087.1 10 1013
          NM_001322159.1 NP_001309088.1 9 951
          NM_001322161.1 NP_001309090.1 9 797
          NM_001322162.1 NP_001309091.1 9 797
          NM_001322164.1 NP_001309093.1 9 816
          NM_001322165.1 NP_001309094.1 8 780
          NM_003205.3 NP_003196.1 9 951
          NM_207036.1 NP_996919.1 9 941
          NM_207037.1 NP_996920.1 9 951
          NM_207038.1 NP_996921.1 9 941
          NM_207040.1 NP_996923.1 2 212
          XM_011521959.2 XP_011520261.1 10 947
          XM_011521960.2 XP_011520262.1 10 947
          XM_011521961.2 XP_011520263.1 10 947
          XM_011521962.2 XP_011520264.1 10 947
          XM_011521963.2 XP_011520265.1 10 947
          XM_011521965.1 XP_011520267.1 9 798
          XM_011521966.2 XP_011520268.1 2 694
          XM_011521967.1 XP_011520269.1 9 798
          XM_011521969.1 XP_011520271.1 2 205
          XM_017022520.1 XP_016878009.1 9 917
          GenBank mRNA AK294617.1 - 9 899
          AK294991.1 - 2 239
          AK302923.1 - 9 901
          AK304007.1 - 2 234
          AK311359.1 - 2 233
          AK312710.1 - 9 911
          AL831981.2 - 9 919
          AU120213.1 - 2 233
          BC050556.1 - 9 892 5767579
          BX537967.1 - 2 203
          M80627.1 - 9 870
          M83233.1 - 9 745

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013176, XM_006243347, XM_006243348, XM_008766219, XM_008766218, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011544, NM_001253862, NM_001253863, NM_001253864, NM_001253865, and others.

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          immune response
          muscle organ development
          positive regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          enhancer binding
          cAMP response element binding
          protein homodimerization activity
          bHLH transcription factor binding
          transcription regulatory region DNA binding
          SMAD binding
          protein heterodimerization activity
          E-box binding
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0