Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116409
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
7
1232
NM_001174093.1
NP_001167564.1
6
892
NM_001174094.1
NP_001167565.1
7
1229
NM_001174095.1
NP_001167566.1
6
751
NM_001174096.1
NP_001167567.1
7
955
NM_001323638.1
NP_001310567.1
11
1602
NM_001323641.1
NP_001310570.1
7
1182
NM_001323642.1
NP_001310571.1
9
1484
NM_001323643.1
NP_001310572.1
9
1262
NM_001323644.1
NP_001310573.1
6
981
NM_001323645.1
NP_001310574.1
8
1325
NM_001323646.1
NP_001310575.1
10
1563
NM_001323647.1
NP_001310576.1
8
1095
NM_001323648.1
NP_001310577.1
7
1035
NM_001323649.1
NP_001310578.1
8
1098
NM_001323650.1
NP_001310579.1
10
1330
NM_001323651.1
NP_001310580.1
8
998
NM_001323652.1
NP_001310581.1
9
1147
NM_001323653.1
NP_001310582.1
9
1254
NM_001323654.1
NP_001310583.1
9
1141
NM_001323655.1
NP_001310584.1
9
1251
NM_001323656.1
NP_001310585.1
7
926
NM_001323657.1
NP_001310586.1
7
929
NM_001323658.1
NP_001310587.1
7
768
NM_001323659.1
NP_001310588.1
7
986
NM_001323660.1
NP_001310589.1
7
983
NM_001323661.1
NP_001310590.1
8
1129
NM_001323662.1
NP_001310591.1
9
1531
NM_001323663.1
NP_001310592.1
10
1470
NM_001323664.1
NP_001310593.1
9
1454
NM_001323665.1
NP_001310594.1
8
1372
NM_001323666.1
NP_001310595.1
7
1312
NM_001323671.1
NP_001310600.1
8
1208
NM_001323672.1
NP_001310601.1
9
1260
NM_001323673.1
NP_001310602.1
8
1205
NM_001323674.1
NP_001310603.1
5
730
NM_001323675.1
NP_001310604.1
5
840
NM_001323676.1
NP_001310605.1
7
1065
NM_001323677.1
NP_001310606.1
7
1062
NM_001323678.1
NP_001310607.1
5
1007
NM_030751.5
NP_110378.3
7
952
XM_006717498.2
XP_006717561.1
10
1390
XM_011519643.2
XP_011517945.1
10
1693
XM_017016597.1
XP_016872086.1
10
1292
XM_017016598.1
XP_016872087.1
10
4422
XM_017016599.1
XP_016872088.1
9
4260
XM_017016600.1
XP_016872089.1
11
4501
XM_017016601.1
XP_016872090.1
10
4419
XM_017016602.1
XP_016872091.1
11
4498
XM_017016603.1
XP_016872092.1
10
4336
GenBank mRNA
AK091478.1
-
6
852
AK294775.1
-
7
1147
AK296244.1
-
7
875
AK299541.1
-
4
489
AK299666.1
-
7
986
AK300830.1
-
6
699
AK307510.1
-
8
958
AK311137.1
-
5
691
AK314683.1
-
7
913
AL831979.1
-
9
1210
BC009572.1
-
7
908
BC065705.1
-
7
913
BC107781.1
-
7
908
BC112392.1
-
7
908
40036600
BX647794.1
-
11
1603
U12170.1
-
7
913
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0