Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATG4B SIRNA Assays ›
          Gene Symbol:
          ATG4B
          Gene Name:
          autophagy related 4B cysteine peptidase
          Gene Aliases:
          APG4B, AUTL1
          Chromosome Location:
          Chr.2: 241637612 - 241673857 on Build GRCh38
          UniGene ID:
          Hs.283610
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 121659
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATG4B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013325.4 NP_037457.3 6 517
          NM_178326.2 NP_847896.1 6 517
          XM_005246992.3 XP_005247049.2 6 503
          XM_005246993.2 XP_005247050.2 6 503
          XM_005246995.3 XP_005247052.1 7 554
          XM_005246996.2 XP_005247053.1 7 499
          XM_017003637.1 XP_016859126.1 6 616
          XM_017003638.1 XP_016859127.1 6 615
          XM_017003639.1 XP_016859128.1 7 573
          XM_017003640.1 XP_016859129.1 8 579
          GenBank mRNA AB023160.2 - 6 424
          AJ504652.1 - 5 420
          AK027332.1 - 6 383
          AK027462.1 - 6 430
          AK027763.1 - 6 450
          AK125277.1 - 6 941
          AK296818.1 - 7 499
          AK298109.1 - 7 527
          AK302963.1 - 7 767
          AK316167.1 - 3 275
          AL080168.1 - 6 518
          BC000719.2 - 6 421 3504267
          BX648007.1 - 7 498

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00367088_m1

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          mitophagy
          C-terminal protein lipidation
          proteolysis
          protein targeting to membrane
          autophagy
          macroautophagy
          protein processing
          nucleophagy
          positive regulation of protein catabolic process
          protein delipidation
          endopeptidase activity
          cysteine-type endopeptidase activity
          protein binding
          cysteine-type peptidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0