Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013281.3 | NP_037413.1 | 2 | 993 | ||
NM_198391.2 | NP_938205.1 | 3 | 1187 | |||
XM_005260682.4 | XP_005260739.1 | 3 | 1705 | |||
XM_011529204.2 | XP_011527506.1 | 4 | 1842 | |||
XM_011529205.2 | XP_011527507.1 | 4 | 1504 | |||
GenBank mRNA | AB040902.1 | - | 2 | 881 | ||
AF169677.1 | - | 2 | 890 | |||
AK027297.1 | - | 2 | 904 | |||
AK027670.1 | - | 1 | 101 | |||
AK027694.1 | - | 1 | 622 | |||
AK074883.1 | - | 2 | 934 | |||
AK074909.1 | - | 2 | 862 | |||
AY358319.1 | - | 1 | 727 | |||
BC020870.1 | - | 2 | 900 | 4606852 |