Search
Search

GALNT18| Gene Symbol: GALNT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198516.2 | NP_940918.2 | 7, 8 | 1707 | ||
| XM_011520068.2 | XP_011518370.1 | 7, 8 | 1283 | |||
| GenBank mRNA | AB078147.1 | - | 7, 8 | 1286 | ||
| AJ626724.1 | - | 7, 8 | 1286 | |||
| AK055111.1 | - | 7, 8 | 1689 | |||
| BC037341.2 | - | 7, 8 | 1696 | 5265475 | ||
| BC060864.1 | - | 7, 8 | 1698 | 30341070 | ||