Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195214.1 | NP_001182143.1 | 6 | 977 | ||
NM_173834.3 | NP_776195.2 | 7 | 1106 | |||
XM_011530924.1 | XP_011529226.1 | 7 | 888 | |||
GenBank mRNA | AK024673.1 | - | 3 | 412 | ||
AK303992.1 | - | 6 | 707 | |||
BC012469.1 | - | 7 | 844 | 4452699 |