Search
Search

SPAG1| Gene Symbol: SPAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003114.4 | NP_003105.2 | 7 | 938 | ||
| NM_172218.2 | NP_757367.1 | 7 | 740 | |||
| XM_011517240.2 | XP_011515542.1 | 7 | 740 | |||
| XM_011517241.2 | XP_011515543.1 | 7 | 740 | |||
| XM_011517242.2 | XP_011515544.1 | 7 | 782 | |||
| XM_011517243.2 | XP_011515545.1 | 7 | 676 | |||
| XM_011517245.2 | XP_011515547.1 | 7 | 740 | |||
| XM_017013754.1 | XP_016869243.1 | 6 | 774 | |||
| XM_017013755.1 | XP_016869244.1 | 6 | 727 | |||
| GenBank mRNA | AF311312.1 | - | 7 | 852 | ||
| AK057482.1 | - | 7 | 740 | |||