Search
Search

ADAMTS16| Gene Symbol: ADAMTS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139056.3 | NP_620687.2 | 7 | 1197 | ||
| NR_136935.1 | 7 | 1197 | ||||
| GenBank mRNA | AB095949.1 | - | 4 | 452 | ||
| AJ315734.1 | - | 7 | 1059 | |||
| AK122980.1 | - | 5 | 822 | |||
| DQ266047.1 | - | 7 | 1197 | |||