Search
Search

SCG2| Gene Symbol: SCG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003469.4 | NP_003460.2 | 2 | 407 | ||
| GenBank mRNA | AK298828.1 | - | 2 | 268 | ||
| AK312452.1 | - | 2 | 268 | |||
| BC022509.1 | - | 2 | 315 | 4820104 | ||
| DA271642.1 | - | 2 | 412 | |||
| M25756.1 | - | 2 | 240 | |||