Search
Search

TRIM54| Gene Symbol: TRIM54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032546.3 | NP_115935.3 | 6 | 1079 | ||
| NM_187841.2 | NP_912730.2 | 5 | 953 | |||
| GenBank mRNA | AK300699.1 | - | 6 | 925 | ||
| BC141807.1 | - | 5 | 725 | 40147443 | ||
| BF829678.1 | - | 2 | 426 | |||