Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006401.2 | NP_006392.1 | 4 | 780 | ||
GenBank mRNA | AK223454.1 | - | 4 | 647 | ||
AK313733.1 | - | 4 | 635 | |||
BC013003.2 | - | 4 | 612 | 3010189 | ||
BC019658.1 | - | 4 | 619 | 4478860 | ||
U70439.1 | - | 4 | 641 | |||
Y07569.1 | - | 4 | 486 | |||
Y07570.1 | - | 4 | 615 | |||
Y07969.1 | - | 4 | 655 |