Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX2 SIRNA Assays ›
Gene Symbol:
RBFOX2
Gene Name:
RNA binding protein, fox-1 homolog 2
Gene Aliases:
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location:
Chr.22: 35738736 - 36028892 on Build GRCh38
UniGene ID:
Hs.282998
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136601
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031695.2
NP_001026865.1
7
1004
NM_001082576.1
NP_001076045.1
7
1004
NM_001082577.1
NP_001076046.1
7
1004
NM_001082578.1
NP_001076047.1
8
964
NM_001082579.1
NP_001076048.1
8
961
NM_014309.2
NP_055124.1
7
1007
XM_005261428.2
XP_005261485.1
8
1319
XM_005261429.2
XP_005261486.1
8
1316
XM_005261430.2
XP_005261487.1
8
1319
XM_005261431.2
XP_005261488.1
8
1319
XM_005261432.2
XP_005261489.1
8
1316
XM_005261433.2
XP_005261490.1
8
1319
XM_005261435.1
XP_005261492.1
7
1008
XM_005261437.1
XP_005261494.1
7
1011
XM_006724185.2
XP_006724248.1
8
1319
XM_006724186.2
XP_006724249.1
8
1316
XM_006724187.2
XP_006724250.1
8
1319
XM_006724188.2
XP_006724251.1
8
1319
XM_006724189.3
XP_006724252.1
8
1012
XM_006724190.1
XP_006724253.1
8
706
XM_006724191.1
XP_006724254.1
7
1011
XM_006724192.2
XP_006724255.1
7
841
XM_006724193.3
XP_006724256.1
8
1319
XM_006724194.2
XP_006724257.1
7
783
XM_017028686.1
XP_016884175.1
8
698
XM_017028687.1
XP_016884176.1
8
879
XM_017028688.1
XP_016884177.1
8
1316
XM_017028689.1
XP_016884178.1
8
784
XM_017028690.1
XP_016884179.1
7
761
XM_017028691.1
XP_016884180.1
7
758
XM_017028692.1
XP_016884181.1
8
1053
XM_017028693.1
XP_016884182.1
7
1011
XM_017028694.1
XP_016884183.1
7
1008
XM_017028695.1
XP_016884184.1
8
768
XM_017028696.1
XP_016884185.1
7
1011
XM_017028697.1
XP_016884186.1
8
1788
XM_017028698.1
XP_016884187.1
7
1008
GenBank mRNA
AB649123.1
-
7
639
AF229058.1
-
7
809
AK055213.1
-
8
925
AK291460.1
-
7
888
AK314632.1
-
7
823
AL009266.1
-
7
812
AM419009.1
-
7
663
AY072786.1
-
8
774
AY960684.1
-
7
639
BC013115.1
-
6
774
4346741
BC025281.1
-
7
827
4555865
CR456559.1
-
7
705
DQ778625.1
-
8
852
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
fibroblast growth factor receptor signaling pathway
regulation of definitive erythrocyte differentiation
RNA metabolic process
radial glia guided migration of Purkinje cell
intracellular estrogen receptor signaling pathway
regulation of cell proliferation
negative regulation of transcription, DNA-templated
dendrite morphogenesis
neuromuscular process controlling balance
nucleotide binding
transcription corepressor activity
RNA binding
mRNA binding
protein binding
transcription factor binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0