Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS6KA5 SIRNA Assays ›
          Gene Symbol:
          RPS6KA5
          Gene Name:
          ribosomal protein S6 kinase A5
          Gene Aliases:
          MSK1, MSPK1, RLPK
          Chromosome Location:
          Chr.14: 90868122 - 91060649 on Build GRCh38
          UniGene ID:
          Hs.510225
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 137676

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPS6KA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322227.1 NP_001309156.1 10, 11 1493
          NM_001322228.1 NP_001309157.1 8, 9 1225
          NM_001322229.1 NP_001309158.1 9, 10 1333
          NM_001322230.1 NP_001309159.1 10, 11 1520
          NM_001322231.1 NP_001309160.1 8, 9 1229
          NM_001322232.1 NP_001309161.1 8, 9 1114
          NM_001322234.1 NP_001309163.1 9, 10 1370
          NM_001322235.1 NP_001309164.1 10, 11 1473
          NM_001322236.1 NP_001309165.1 8, 9 1249
          NM_001322237.1 NP_001309166.1 9, 10 1132
          NM_001322238.1 NP_001309167.1 7, 8 841
          NM_004755.3 NP_004746.2 9, 10 1333
          NM_182398.2 NP_872198.1 9, 10 1333
          XM_017021786.1 XP_016877275.1 10, 11 1372
          GenBank mRNA AF074393.1 - 9, 10 1283
          AF080000.1 - 9, 10 1299
          AF090421.1 - 9, 10 1282
          AK295266.1 - 9, 10 1132
          AK309925.1 - 9, 10 1357
          BC017187.2 - 9, 10 1284 3139114

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cytokine production
          stimulatory C-type lectin receptor signaling pathway
          regulation of transcription, DNA-templated
          protein phosphorylation
          inflammatory response
          epidermal growth factor receptor signaling pathway
          axon guidance
          histone phosphorylation
          positive regulation of CREB transcription factor activity
          positive regulation of histone phosphorylation
          positive regulation of histone acetylation
          intracellular signal transduction
          histone H3-S10 phosphorylation
          histone H3-S28 phosphorylation
          histone H2A-S1 phosphorylation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of NF-kappaB transcription factor activity
          interleukin-1-mediated signaling pathway
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          histone kinase activity (H3-S10 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0