Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PROM1 SIRNA Assays ›
Gene Symbol:
PROM1
Gene Name:
prominin 1
Gene Aliases:
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
Chromosome Location:
Chr.4: 15968226 - 16084100 on Build GRCh38
UniGene ID:
Hs.614734
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138146
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PROM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145847.1
NP_001139319.1
5
1046
NM_001145848.1
NP_001139320.1
5
762
NM_001145849.1
NP_001139321.1
5
766
NM_001145850.1
NP_001139322.1
5
766
NM_001145851.1
NP_001139323.1
4
739
NM_001145852.1
NP_001139324.1
4
739
NM_006017.2
NP_006008.1
5
766
XM_005248195.4
XP_005248252.1
5
862
XM_005248196.4
XP_005248253.1
5
862
XM_006713974.3
XP_006714037.1
4
367
XM_011513890.1
XP_011512192.1
6
848
XM_011513892.2
XP_011512194.1
6
999
XM_011513893.2
XP_011512195.1
6
1073
XM_011513894.2
XP_011512196.1
6
1158
XM_011513895.2
XP_011512197.1
7
1274
XM_011513896.2
XP_011512198.1
6
903
XM_011513897.2
XP_011512199.1
6
1090
XM_011513898.2
XP_011512200.2
6
848
XM_011513899.2
XP_011512201.2
5
862
XM_011513900.2
XP_011512202.1
6
848
XM_011513902.2
XP_011512204.1
6
848
XM_011513903.2
XP_011512205.1
5
397
XM_017008799.1
XP_016864288.1
5
892
XM_017008800.1
XP_016864289.1
6
848
XM_017008801.1
XP_016864290.1
6
848
XM_017008802.1
XP_016864291.1
6
940
XM_017008803.1
XP_016864292.1
5
837
XM_017008804.1
XP_016864293.1
5
907
XM_017008805.1
XP_016864294.1
5
780
GenBank mRNA
AF027208.1
-
5
591
AF117225.1
-
5
807
AF507034.1
-
4
564
AK027420.1
-
5
785
AK027422.1
-
5
807
AK075411.1
-
5
762
AK226033.1
-
5
785
AK310415.1
-
5
612
AK314529.1
-
5
807
AK315889.1
-
2
195
AY449689.1
-
4
527
AY449690.1
-
4
527
AY449691.1
-
4
527
AY449692.1
-
5
554
AY449693.1
-
5
554
BC012089.1
-
5
762
4644690
HQ628626.1
-
4
527
HQ628627.1
-
6
783
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
retina layer formation
photoreceptor cell maintenance
retina morphogenesis in camera-type eye
camera-type eye photoreceptor cell differentiation
glomerular visceral epithelial cell differentiation
glomerular parietal epithelial cell differentiation
positive regulation of nephron tubule epithelial cell differentiation
protein binding
actinin binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0