Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDCD6IP SIRNA Assays ›
          Gene Symbol:
          PDCD6IP
          Gene Name:
          programmed cell death 6 interacting protein
          Gene Aliases:
          AIP1, ALIX, DRIP4, HP95
          Chromosome Location:
          Chr.3: 33798571 - 33869707 on Build GRCh38
          UniGene ID:
          Hs.475896
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138252

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDCD6IP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001162429.2 NP_001155901.1 11 1614
          NM_013374.5 NP_037506.2 11 1599
          XM_011533252.1 XP_011531554.1 11 1837
          XM_011533253.1 XP_011531555.1 11 1376
          XM_017005488.1 XP_016860977.1 11 1361
          GenBank mRNA AB037796.1 - 4 475
          AF151793.1 - 11 1567
          AF250394.1 - 11 1531
          AF349951.1 - 11 1572
          AK002122.1 - 6 819
          AK295042.1 - 7 946
          AK298727.1 - 5 940
          BC020066.1 - 11 1549 4340998
          BC068454.1 - 11 1595 4830724
          BT007367.1 - 11 1441

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell separation after cytokinesis
          apoptotic process
          nucleus organization
          mitotic metaphase plate congression
          regulation of centrosome duplication
          protein transport
          viral life cycle
          multivesicular body assembly
          viral budding via host ESCRT complex
          ubiquitin-independent protein catabolic process via the multivesicular body sorting pathway
          regulation of mitotic spindle assembly
          positive regulation of exosomal secretion
          regulation of extracellular exosome assembly
          positive regulation of extracellular exosome assembly
          protein binding
          proteinase activated receptor binding
          protein homodimerization activity
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fxx5q:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0