Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BEST1 SIRNA Assays ›
          Gene Symbol:
          BEST1
          Gene Name:
          bestrophin 1
          Gene Aliases:
          ARB, BEST, BMD, RP50, TU15B, VMD2
          Chromosome Location:
          Chr.11: 61946722 - 61964465 on Build GRCh38
          UniGene ID:
          Hs.524910
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 139364

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BEST1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001139443.1 NP_001132915.1 8 1399
          NM_001300786.1 NP_001287715.1 7 1318
          NM_001300787.1 NP_001287716.1 8 1399
          NM_004183.3 NP_004174.1 9 1587
          NR_134580.1 9 1790
          XM_005274210.3 XP_005274267.1 9 1697
          XM_005274215.3 XP_005274272.1 7 1352
          XM_005274216.3 XP_005274273.1 8 1712
          XM_011545229.2 XP_011543531.1 9 1137
          XM_011545230.2 XP_011543532.1 8 940
          XM_011545233.2 XP_011543535.1 3 834
          XM_017018228.1 XP_016873717.1 7 1352
          XM_017018229.1 XP_016873718.1 9 1900
          XM_017018230.1 XP_016873719.1 7 1555
          GenBank mRNA AB209451.1 - 6 2628
          AF057169.1 - 9 1111
          AF057170.1 - 9 1314
          AF073501.1 - 9 1095
          AK289681.1 - 9 1318
          AK293701.1 - 8 1053
          AK295450.1 - 8 1133
          AK295998.1 - 8 933
          AK301392.1 - 7 916
          AY515704.1 - 8 1007
          BC015220.2 - 7 1318 3877806
          BC041664.1 - 8 913 5194649
          JQ954696.1 - 8 1073
          KU178390.1 - 6 746
          KU178391.1 - 7 827
          KU178392.1 - 6 674

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          chloride transport
          visual perception
          bicarbonate transport
          transepithelial chloride transport
          ion transmembrane transport
          detection of light stimulus involved in visual perception
          regulation of calcium ion transport
          chloride transmembrane transport
          intracellular calcium activated chloride channel activity
          chloride channel activity
          bicarbonate transmembrane transporter activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0