Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291333.1 | NP_001278262.1 | 3 | 387 | ||
NM_004623.4 | NP_004614.3 | 3 | 387 | |||
GenBank mRNA | AF063602.1 | - | 3 | 349 | ||
AF073887.1 | - | 3 | 333 | |||
AK025524.1 | - | 3 | 387 | |||
AK090964.1 | - | 3 | 467 | |||
AK222562.1 | - | 3 | 340 | |||
AK292426.1 | - | 3 | 348 | |||
BC001276.1 | - | 3 | 326 | 3462738 | ||
BT006817.1 | - | 3 | 300 |
Gene Symbol: MROH7-TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037639.2 | 26 | 4478 | |||
NR_037640.2 | 26 | 4345 | ||||
NR_037641.2 | 25 | 4229 | ||||
GenBank mRNA | AK127008.1 | - | 10 | 1979 | ||
BX648088.1 | - | 26 | 4284 | |||
CR749830.1 | - | 26 | 4430 |