Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BANP SIRNA Assays ›
          Gene Symbol:
          BANP RRN3P2
          Gene Name:
          BTG3 associated nuclear protein
          RRN3 homolog, RNA polymerase I transcription factor pseudogene 2
          Gene Aliases:
          BEND1, SMAR1, SMARBP1
          -
          Chromosome Location:
          Chr.16: 87949216 - 88077318 on Build GRCh38
          Chromosome Location:
          Chr.16: 29074842 - 29116718 on Build GRCh38
          UniGene ID:
          Hs.461705
          Hs.720348
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 140456

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BANP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001173539.1 NP_001167010.1 3 286
          NM_001173540.1 NP_001167011.1 3 304
          NM_001173541.1 NP_001167012.1 3 286
          NM_001173542.1 NP_001167013.1 3 325
          NM_001173543.1 NP_001167014.1 3 355
          NM_017869.3 NP_060339.2 3 286
          NM_079837.2 NP_524576.2 3 286
          XM_005256008.3 XP_005256065.1 3 302
          XM_005256009.3 XP_005256066.1 3 284
          XM_011523170.2 XP_011521472.1 3 302
          XM_011523172.2 XP_011521474.1 4 547
          XM_011523174.2 XP_011521476.1 3 1073
          XM_011523175.2 XP_011521477.1 3 704
          XM_011523176.2 XP_011521478.1 3 766
          XM_011523177.2 XP_011521479.1 3 1274
          XM_011523178.2 XP_011521480.1 3 302
          XM_011523179.2 XP_011521481.1 3 284
          XM_011523180.2 XP_011521482.1 4 366
          XM_011523181.2 XP_011521483.1 3 302
          XM_011523182.1 XP_011521484.1 3 302
          XM_011523183.2 XP_011521485.1 3 279
          XM_011523184.2 XP_011521486.1 3 235
          XM_011523186.2 XP_011521488.1 1 64
          XM_017023378.1 XP_016878867.1 4 406
          XM_017023379.1 XP_016878868.1 3 686
          XM_017023380.1 XP_016878869.1 3 286
          XM_017023381.1 XP_016878870.1 3 304
          XM_017023383.1 XP_016878872.1 3 251
          GenBank mRNA AK001039.1 - 3 286
          AK094158.1 - 3 356
          AK125232.1 - 3 284
          AK293472.1 - 3 248
          AK297945.1 - 3 258
          AK315089.1 - 3 284
          AK315535.1 - 3 325
          BC009424.1 - 3 231 3530821
          DA632793.1 - 3 286
          KU178664.1 - 2 134
          KU178665.1 - 2 134
          KU178666.1 - 2 134
          KU178667.1 - 2 134
          KU178668.1 - 2 134
          KU178669.1 - 2 134
          KU178670.1 - 2 156
          Gene Symbol: RRN3P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003369.2 16 1777
          GenBank mRNA AK225670.1 - 16 1627
          AK298442.1 - 14 1518

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          cell cycle
          multicellular organism development
          covalent chromatin modification
          protein localization to nucleus
          negative regulation of protein catabolic process
          positive regulation of transcription, DNA-templated
          regulation of signal transduction by p53 class mediator
          p53 binding
          DNA binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0