Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022070.4 | NP_071353.4 | 11 | 1676 | ||
XM_005257570.3 | XP_005257627.1 | 12 | 1864 | |||
XM_011525110.2 | XP_011523412.1 | 10 | 1563 | |||
XM_011525111.2 | XP_011523413.1 | 10 | 1525 | |||
XM_017024936.1 | XP_016880425.1 | 11 | 1937 | |||
XM_017024937.1 | XP_016880426.1 | 11 | 3613 | |||
XM_017024938.1 | XP_016880427.1 | 11 | 1669 | |||
XM_017024939.1 | XP_016880428.1 | 11 | 1940 | |||
GenBank mRNA | AF349752.1 | - | 12 | 1621 | ||
AK025740.1 | - | 4 | 743 | |||
AK026070.1 | - | 4 | 516 | |||
AK127708.1 | - | 11 | 1670 | |||
AK314655.1 | - | 11 | 1679 | |||
BC119771.1 | - | 11 | 1666 | 40108435 | ||
BC119813.1 | - | 11 | 1667 | 40108426 | ||
BX640819.1 | - | 11 | 1667 | |||
BX640831.1 | - | 11 | 1667 | |||
CR627418.1 | - | 11 | 1666 |