Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLTM SIRNA Assays ›
Gene Symbol:
SLTM
Gene Name:
SAFB like transcription modulator
Gene Aliases:
Met
Chromosome Location:
Chr.15: 58879045 - 58934313 on Build GRCh38
UniGene ID:
Hs.512932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140877
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLTM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013843.2
NP_001013865.1
15
1997
NM_024755.3
NP_079031.2
15
2051
NR_135042.1
13
1384
NR_135043.1
13
1494
XM_006720686.3
XP_006720749.3
13
2442
XM_006720690.1
XP_006720753.1
11
1148
XM_011522022.1
XP_011520324.1
15
2709
XM_011522023.1
XP_011520325.1
15
2655
XM_011522024.1
XP_011520326.1
14
2559
XM_011522025.1
XP_011520327.1
14
2511
XM_011522026.2
XP_011520328.2
14
2490
XM_011522027.1
XP_011520329.1
13
2463
XM_011522028.1
XP_011520330.1
13
2409
XM_011522029.2
XP_011520331.1
15
1890
XM_011522030.2
XP_011520332.1
15
2709
XM_011522031.2
XP_011520333.1
12
1545
XM_017022576.1
XP_016878065.1
14
1847
XM_017022577.1
XP_016878066.1
14
2436
XM_017022578.1
XP_016878067.1
13
2388
XM_017022579.1
XP_016878068.1
15
2054
XM_017022580.1
XP_016878069.1
14
1508
GenBank mRNA
AK023275.1
-
5
505
AK055195.1
-
13
1287
AK291423.1
-
13
1417
BC014944.1
-
8
854
BC046119.1
-
4
346
BC140851.1
-
15
2018
9021719
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-97z6t:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0