Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKACA SIRNA Assays ›
          Gene Symbol:
          PRKACA
          Gene Name:
          protein kinase cAMP-activated catalytic subunit alpha
          Gene Aliases:
          PKACA, PPNAD4
          Chromosome Location:
          Chr.19: 14091688 - 14117747 on Build GRCh38
          UniGene ID:
          Hs.631630
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 142274

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKACA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001304349.1 NP_001291278.1 6 1103
          NM_002730.3 NP_002721.1 6 669
          NM_207518.2 NP_997401.1 6 478
          XM_017026948.1 XP_016882437.1 6 525
          GenBank mRNA AF208004.1 - 6 463
          AK093803.1 - 1 625
          AK131561.1 - 6 1103
          AK290147.1 - 7 551
          AK292284.1 - 6 478
          AK301264.1 - 6 838
          BC039846.1 - 6 665 6066764
          BC108259.1 - 6 666 6144307
          BU543573.1 - 669
          M80335.1 - 5 418
          X07767.1 - 6 549

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_008854, NM_001277898, CD807077, BC003238, BC054834, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          mesoderm formation
          neural tube closure
          regulation of heart rate
          stimulatory C-type lectin receptor signaling pathway
          renal water homeostasis
          mRNA processing
          protein phosphorylation
          blood coagulation
          positive regulation of cell proliferation
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          activation of protein kinase A activity
          calcium-mediated signaling using intracellular calcium source
          lipoprotein metabolic process
          regulation of protein binding
          regulation of cellular respiration
          regulation of osteoblast differentiation
          protein autophosphorylation
          positive regulation of protein export from nucleus
          sperm capacitation
          negative regulation of meiotic cell cycle
          regulation of cytosolic calcium ion concentration
          regulation of synaptic transmission, glutamatergic
          regulation of cardiac muscle contraction
          regulation of ryanodine-sensitive calcium-release channel activity
          regulation of proteasomal protein catabolic process
          regulation of protein processing
          positive regulation of cell cycle arrest
          cellular response to glucose stimulus
          cellular response to parathyroid hormone stimulus
          cellular response to glucagon stimulus
          cellular response to epinephrine stimulus
          cell communication by electrical coupling involved in cardiac conduction
          negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          regulation of cardiac conduction
          regulation of bicellular tight junction assembly
          protein serine/threonine kinase activity
          cAMP-dependent protein kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          protein kinase binding
          ubiquitin protein ligase binding
          protein complex binding
          protein kinase A regulatory subunit binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0