Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIPC SIRNA Assays ›
          Gene Symbol:
          LIPC
          Gene Name:
          lipase C, hepatic type
          Gene Aliases:
          HDLCQ12, HL, HTGL, LIPH
          Chromosome Location:
          Chr.15: 58410754 - 58568952 on Build GRCh38
          UniGene ID:
          Hs.654472
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143213

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LIPC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000236.2 NP_000227.2 4 544
          XM_005254372.1 XP_005254429.1 5 592
          XM_005254374.4 XP_005254431.2 4 1886
          XM_006720502.3 XP_006720565.1 4 489
          XM_017022176.1 XP_016877665.1 4 1901
          GenBank mRNA AK290062.1 - 6 1095
          AK292631.1 - 6 1098
          AK293324.1 - 3 346
          AK309448.1 - 3 344
          AK315306.1 - 4 723
          BC132825.1 - 4 522 40146847
          BC136495.1 - 4 522 9020484
          BC146659.1 - 5 767 40148028
          D83548.1 - 4 528
          J03540.1 - 4 491
          J03895.1 - 4 494
          X07228.1 - 4 544

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lipase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          liver development
          fatty acid biosynthetic process
          phosphatidylserine metabolic process
          circadian rhythm
          cholesterol metabolic process
          response to carbohydrate
          response to acetate
          heparan sulfate proteoglycan biosynthetic process
          triglyceride catabolic process
          response to nutrient levels
          response to magnesium ion
          cellular response to hormone stimulus
          chylomicron remodeling
          very-low-density lipoprotein particle remodeling
          intermediate-density lipoprotein particle remodeling
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          chylomicron remnant clearance
          low-density lipoprotein particle clearance
          phosphatidylcholine catabolic process
          response to drug
          cholesterol homeostasis
          response to amino acid
          response to peptide hormone
          reverse cholesterol transport
          lipid digestion
          phosphatidylethanolamine metabolic process
          phosphatidic acid metabolic process
          glycerophospholipid catabolic process
          response to copper ion
          developmental growth
          protein oligomerization
          response to glucocorticoid
          response to calcium ion
          triglyceride homeostasis
          response to fatty acid
          phospholipase activity
          lysophospholipase activity
          triglyceride lipase activity
          heparin binding
          lipid binding
          phosphatidylcholine 1-acylhydrolase activity
          transferase activity, transferring acyl groups
          low-density lipoprotein particle binding
          apolipoprotein binding
          heparan sulfate proteoglycan binding
          acylglycerol lipase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0