Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LCK SIRNA Assays ›
          Gene Symbol:
          LCK
          Gene Name:
          LCK proto-oncogene, Src family tyrosine kinase
          Gene Aliases:
          IMD22, LSK, YT16, p56lck, pp58lck
          Chromosome Location:
          Chr.1: 32251239 - 32286167 on Build GRCh38
          UniGene ID:
          Hs.470627
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143991

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LCK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042771.2 NP_001036236.1 12 1360
          NM_005356.4 NP_005347.3 12 1377
          XM_011541453.2 XP_011539755.1 10 1398
          XM_011541454.2 XP_011539756.1 10 1379
          XM_011541455.2 XP_011539757.1 11 1224
          GenBank mRNA AF228313.1 - 11 1200
          AJ865079.1 - 9 1260
          AK098027.1 - 12 1174
          AK303073.1 - 11 1430
          BC013200.1 - 12 1429 4341278
          KU178161.1 - 11 1329
          KU178162.1 - 11 1239
          KU178163.1 - 10 1086
          M36881.1 - 12 1290
          U07236.1 - 12 1274
          X04476.1 - 2 117
          X05027.1 - 12 1334
          X06369.1 - 2 138
          X13529.1 - 12 1290

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          protein dephosphorylation
          cellular zinc ion homeostasis
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          transmembrane receptor protein tyrosine kinase signaling pathway
          regulation of phosphatidylinositol 3-kinase signaling
          viral process
          hemopoiesis
          platelet activation
          T cell differentiation
          T cell costimulation
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          response to drug
          innate immune response
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          regulation of defense response to virus by virus
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          positive regulation of T cell receptor signaling pathway
          positive regulation of T cell activation
          leukocyte migration
          release of sequestered calcium ion into cytosol
          regulation of lymphocyte activation
          positive regulation of intrinsic apoptotic signaling pathway
          glycoprotein binding
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          protein serine/threonine phosphatase activity
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          protein kinase binding
          protein phosphatase binding
          SH2 domain binding
          CD4 receptor binding
          CD8 receptor binding
          identical protein binding
          phosphatidylinositol 3-kinase binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0