Search
Search

MTNR1A| Gene Symbol: MTNR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005958.4 | NP_005949.1 | 2 | 1188 | ||
| XM_011532002.2 | XP_011530304.1 | 2 | 1269 | |||
| GenBank mRNA | BC074946.2 | - | 2 | 1004 | 30915497 | |
| BC074947.2 | - | 2 | 1004 | 30915274 | ||
| BC113816.1 | - | 1 | 639 | |||
| BC126297.1 | - | 2 | 1188 | 8992013 | ||
| BC126299.1 | - | 2 | 1186 | 8992015 | ||
| EU432127.1 | - | 2 | 986 | |||
| U14108.1 | - | 2 | 1018 | |||
Set Membership: |
|